Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human Collagen Type I ELISA Kit (Colorimetric)
SDP

Catalog No. NBP230102 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP230102 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP230102 Supplier Novus Biologicals™ Supplier No. NBP230102
Add to Cart
Add to Cart

ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.

The Human Collagen Type I ELISA Kit (Colorimetric) measures human Collagen Type I in serum, plasma and other biological fluids.

Specifications

Assay Range 0.156 to 10 ng/mL (example only; lot dependent)
Assay Sensitivity 0.229ng/mL
Product Type ELISA Kit
Target Species Human
For Use With (Application) ELISA
Kit Contents Microtiter Plate, 1x96wells, Pre-coated, ready to use, Standard, 2x1vial, Standard Diluent, 1x20ml, Detection Reagent A, 1x120ul, Detection Reagent B, 1x120ul, Assay Diluent A, 2x, 1x6ml, Assay Diluent B, 2x, 1x6ml, TMB Substrate, 1x9ml, Positive Control, 4.74ng/ml, 1x1vial, Stop Solution, 1x6ml, Wash Buffer, 30x, 1x20ml
Quantity 1 Kit
Storage Requirements Storage is content dependent.
Synonym Collagen I, Collagen Type 1
Target Collagen Type I
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.