Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Collagen XXIII alpha 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF4165
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF4165 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF4165 Supplier R&D Systems Supplier No. AF4165
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Collagen XXIII alpha 1 Polyclonal specifically detects Collagen XXIII alpha 1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Collagen XXIII alpha 1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q86Y22
Gene Alias COL23A1, collagen alpha-1(XXIII) chain, collagen, type XXIII, alpha 1, DKFZp434K0621, procollagen, type XXIII, alpha 1
Gene Symbols COL23A1
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Collagen XXIII alpha 1 Glu111-Lys540 Accession # Q86Y22
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91522
Test Specificity Detects human Collagen XXIII alpha 1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human COL25A1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.