Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Common beta Chain Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF906SP
missing translation for 'nonReturnableMessage'
missing translation for 'returnPolicyLabel'
Description
Common beta Chain Polyclonal specifically detects Common beta Chain in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
Common beta Chain | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
beta, low-affinity, CD131, CD131 antigen, CDw131, colony stimulating factor 2 receptor, beta, low-affinity(granulocyte-macrophage), CSF2RB, cytokine receptor common subunit beta, GM-CSF R beta, GM-CSF/IL-3/IL-5 receptor common beta subunit, GM-CSF/IL-3/IL-5 receptor common beta-chain, IL-3 R beta, IL3RB, IL-5 R beta, interleukin 3 receptor/granulocyte-macrophage colony stimulating factor 3receptor, beta (high affinity) | |
Goat | |
Affinity Purified | |
RUO | |
1439 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
P32927 | |
CSF2RB | |
Mouse myeloma cell line NS0-derived recombinant human Common beta Chain Gly23-Trp443 Accession # P32927 | |
25 μg | |
Primary | |
Detects recombinant human (rh) Common beta Chain in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) GM-CSF R alpha , rhG-CSF R and rhM-CSF R is observed. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction