Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Common gamma Chain/IL-2 R gamma DuoSet ELISA
SDP

Catalog No. DY28405 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit (for 5 Plates)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DY28405 1 Kit (for 5 Plates)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DY28405 Supplier R&D Systems™ Supplier No. DY28405
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Capture antibody, detection antibody, recombinant standard, streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols

For the determination of natural and recombinant common gamma chain in humans. R&D Systems Human Common gamma Chain/IL-2 R gamma DuoSet ELISA Kit is a solid phase sandwich ELISA suitable for the analysis of cell culture supernate, serum, and plasma samples.

The Human Common gamma Chain/IL-2 R gamma DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human Common gamma Chain/IL-2 R gamma.

Specifications

Description Common gamma Chain/IL-2 R gamma
Product Type ELISA Kit
Quantity 1 Kit (for 5 Plates)
Species Human
Cross Reactivity Not observed
Format 96-well Strip Plate
For Use With (Application) ELISA
Sample Type Cell Culture Supernatant, Serum, Plasma
Kit Contents Capture antibody, Detection antibody, Recombinant standard, Streptavidin conjugated to horseradish-peroxidase (Streptavidin-HRP) and development protocols
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.