Learn More
Specifications
Specifications
| Assay Range | 1.56-100 ng/mL |
| Assay Sensitivity | 0.94 ng/mL |
| Product Type | ELISA Kit (Colorimetric) |
| Sample Type | Biological Fluids, Plasma, Serum |
| Specificity | This kit recognizes Human Complement C4b in samples. No significant cross-reactivity or interference between Human Complement C4b and analogues was observed. |
| For Use With (Application) | ELISA, Sandwich ELISA |
| Gene ID (Entrez) | 721 |
| Gene Symbol | C4B |
| Quantity | 1 Kit |
| Storage Requirements | Storage of components varies. See protocol for specific instructions. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
