Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Complement Component C1r Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1807
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1807 100 μg
MAB1807SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1807 Supplier R&D Systems Supplier No. MAB1807
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement Component C1r Monoclonal specifically detects Complement Component C1r in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Complement Component C1r
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 269104
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_001724
Gene Alias C1r, complement C1r subcomponent, Complement component 1 subcomponent r, complement component 1, r subcomponent, EC 3.4.21, EC 3.4.21.41
Gene Symbols C1R
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Complement Component C1r Ser18-Asp705 Accession # NP_001724
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 715
Test Specificity Detects human C1r in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) C1s or rhC1q R1 is observed. In Western blots, ths antibody recognizes rhC1r and the A/heavy chain.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.