Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Complement Component C1rLP Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | Complement Component C1rLP |
---|---|
Clone | 393608 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Complement Component C1rLP Monoclonal specifically detects Complement Component C1rLP in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Complement Component C1rLP | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
AAI56781 | |
51279 | |
Mouse myeloma cell line NS0-derived recombinant human Complement Component C1rLP Ser41-Asn487 Accession # AAI56781 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Complement Component C1rLP in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) C1r, rhC1s, rhHaptoglobin, or rhMASP3 is observed. |
393608 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C1RL, C1RL1, C1RL1complement C1r subcomponent-like protein, C1r-like protein, C1r-like serine protease analog protein, C1r-LPcomplement C1r-like proteinase, C1RLPEC 3.4.21.-, CLSPa, complement component 1, r subcomponent-like, EC 3.4.21, EC 3.4.21.41 | |
C1RL | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title