Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Complement Component C1s Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | Complement Component C1s |
---|---|
Clone | 306904 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Description
Complement Component C1s Monoclonal specifically detects Complement Component C1s in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Complement Component C1s | |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human | |
P09871 | |
716 | |
Mouse myeloma cell line NS0-derived recombinant human Complement Component C1s Glu16-Asp688 Accession # P09871 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human Complement Component C1s in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human C1r is observed. |
306904 | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
basic proline-rich peptide IB-1, C1 esterase, C1S, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757 | |
C1S | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title