Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Complement Component C1s Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAF2060
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAF2060 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAF2060 Supplier R&D Systems Supplier No. BAF2060
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Complement Component C1s Polyclonal specifically detects Complement Component C1s in Human samples. It is validated for Western Blot.

Specifications

Antigen Complement Component C1s
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Biotin
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P09871
Gene Alias basic proline-rich peptide IB-1, C1 esterase, C1S, complement C1s subcomponent, Complement component 1 subcomponent s, complement component 1, s subcomponent, EC 3.4.21, EC 3.4.21.42, FLJ44757
Gene Symbols C1S
Host Species Sheep
Immunogen Mouse myeloma cell line NS0-derived recombinant human Complement Component C1s Glu16-Asp688 Accession # P09871
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 716
Test Specificity Detects human Complement Component C1s in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human C1r is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.