Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Complement Component C2b Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB1936SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1936SP 25 μg
MAB1936 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB1936SP Supplier R&D Systems Supplier No. MAB1936SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement Component C2b Monoclonal specifically detects Complement Component C2b in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Complement Component C2b
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 269716
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P06681
Gene Alias ARMD14, CO2
Gene Symbols C2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Complement Component C2 Ala21-Leu752 Accession # P06681
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 717
Test Specificity Detects human Complement Component C2b in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) C1r, rhC1s, rhC3r, rhC5a, rhFactor B, or rhFactor I is observed. In Western blots, this antibody recognizes rhC2 and rhC2B.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.