Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Complement Component C3a Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF3677
This item is not returnable.
View return policy
Description
Complement Component C3a Polyclonal specifically detects Complement Component C3a in Human samples. It is validated for Western Blot, Immunocytochemistry.Specifications
Complement Component C3a | |
Polyclonal | |
Unconjugated | |
P01024 | |
Goat | |
Affinity Purified | |
RUO | |
718 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
C3 | |
E. coli-derived recombinant human C3a Ser672-Arg748 Accession # P01024 | |
100 μg | |
Primary | |
Detects human Complement Component C3a in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction