Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Complement Component C5/C5a Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. MAB2037SP Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2037SP 25 μg
MAB2037100 100 μg
MAB2037500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB2037SP Supplier Novus Biologicals Supplier No. MAB2037SP
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

Complement Component C5/C5a Monoclonal specifically detects Complement Component C5/C5a in Human∣Rat samples. It is validated for Bioassay∣ELISA Capture∣ELISA Capture (Matched Antibody Pair)∣ELISA Development∣ELISA Development (Capture)∣Neutralization∣Western Blot.

Specifications

Antigen Complement Component C5/C5a
Applications Bioassay, ELISA Capture (Matched Antibody Pair), ELISA Capture (Matched Antibody Pair), ELISA Development, ELISA Development (Capture), Neutralization, Western Blot
Classification Monoclonal
Clone 295003
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Neutralization 0.04-0.12 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P01031
Gene Symbols C5
Host Species Mouse
Immunogen E. coli-derived recombinant human Complement Component C5a Thr678-Arg751 Accession # P01031
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727
Test Specificity Detects human Complement Component C5/C5a in ELISAs and Western blots. In sandwich immunoassays, detects human Complement Component C5a by itself or in the context of Complement Component C5. In Western blots, this antibody does not cross-react with recombinant human Complement Component C2, recombinant mouse (rm) Complement Component C3d, or rmComplement Component C5a.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Rat
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.