Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Complement Component C5/C5a Biotinylated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. BAM20371
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BAM20371 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. BAM20371 Supplier R&D Systems Supplier No. BAM20371
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

Complement Component C5/C5a Monoclonal specifically detects Complement Component C5/C5a in Human samples. It is validated for ELISA Detection (Matched Antibody Pair).

Specifications

Antigen Complement Component C5/C5a
Applications ELISA
Classification Monoclonal
Clone 295009
Concentration LYOPH
Conjugate Biotin
Dilution ELISA Detection (Matched Antibody Pair) 0.5-2.0 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. with No Preservative
Gene Accession No. P01031
Gene Symbols C5
Host Species Mouse
Immunogen E. coli-derived recombinant human Complement Component C5a Thr678-Arg751 Accession # P01031
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 727
Test Specificity Detects human Complement Component C5/C5a in ELISAs. In sandwich immunoassays, detects human Complement Component C5a by itself or in the context of Complement Component C5. In ELISAs, this antibody does not cross-react with recombinant human (rh) Complement Component C3a, rh alpha 2-macroglobulin, recombinant mouse (rm) Complement Component C5a, or rmComplement Component C5d.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.