Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Complement Component C5aR1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB36481U100UG
This item is not returnable.
View return policy
Description
Complement Component C5aR1 Monoclonal specifically detects Complement Component C5aR1 in Human samples. It is validated for Flow Cytometry.Specifications
Complement Component C5aR1 | |
Monoclonal | |
Alexa Fluor 350 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
C5A, C5a anaphylatoxin chemotactic receptor, C5aR, C5a-R, C5AR1, C5ARC5a anaphylatoxin receptor, C5R1C5a ligand, CD88, CD88 antigen, complement component 5 receptor 1, complement component 5 receptor 1 (C5a ligand), complement component 5a receptor 1, Complement Component C5a R1 | |
NS0 mouse myeloma cell line transfected with human Complement Component C5aR1, Met1-Val350, Accession # P21730 | |
100 μg | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
347234 | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P21730 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
728 | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction