Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Complement Component C9 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF8126SP
This item is not returnable.
View return policy
Description
Complement Component C9 Polyclonal specifically detects Complement Component C9 in Human samples. It is validated for Western Blot.Specifications
| Complement Component C9 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| complement component 9, complement component C9 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 735 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.2 ug/mL | |
| P02748 | |
| C9 | |
| Human embryonic kidney cell line HEK293-derived human Complement Component C9 Gln22-Lys559 (predicted) Accession # P02748 | |
| 25 μg | |
| Primary | |
| Detects human Complement Component C9 in directs ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction