Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Complement Factor H-related 2/CFHR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5484SP
This item is not returnable.
View return policy
Description
Complement Factor H-related 2/CFHR2/CFHL2 Monoclonal specifically detects Complement Factor H-related 2/CFHR2/CFHL2 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Complement Factor H-related 2/CFHR2/CFHL2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
P36980 | |
CFHR2 | |
Mouse myeloma cell line NS0-derived recombinant human Complement Factor H-related 2/CFHR2 Glu19-Glu268 Accession # P36980 | |
25 μg | |
Primary | |
Detects human Complement Factor H-related 2/CFHR2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CFHR1 or rhCFHR5 is observed. | |
Human | |
Purified |
Western Blot, Immunoprecipitation | |
510511 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CFHL2, CFHL2FHR-2, CFHR2, complement factor H-related 2, Complement Factor Hrelated 2, factor H-related gene 2, FHR2, FHR2DDESK59, H factor (complement)-like 3, H factor-like 3, H factor-like protein 2, HFL3, HFL3complement factor H-related protein 2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3080 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction