Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Complement Factor H-related 4/CFHR4 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC5980V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC5980V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC5980V100 Supplier R&D Systems Supplier No. IC5980V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement Factor H-related 4/CFHR4 Monoclonal specifically detects Complement Factor H-related 4/CFHR4 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Complement Factor H-related 4/CFHR4
Applications Flow Cytometry
Classification Monoclonal
Clone 640212
Conjugate Alexa Fluor 405
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q92496
Gene Alias complement factor H-related protein 4, CFHL4, CFHR4, complement factor H-related 4, Complement Factor Hrelated 4, FHR4, FHR-4
Gene Symbols CFHR4
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Complement Factor H-related 4/CFHR4 Glu20-Glu331 Accession # Q92496
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10877
Test Specificity Detects human Complement Factor H-related 4/CFHR4 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) CFHR1, rhCFHR2, or rhCFHR5 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.