Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Complement Factor H-related 5/ CFHR5 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3845SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3845SP 25 μg
MAB3845 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3845SP Supplier R&D Systems Supplier No. MAB3845SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Complement Factor H-related 5/CFHR5 Monoclonal specifically detects Complement Factor H-related 5/CFHR5 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen Complement Factor H-related 5/CFHR5
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone 390513
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9BXR6
Gene Alias CFHL5, CFHL5factor H-related protein 5, CFHR5, complement factor H-related 5, Complement Factor Hrelated 5, FHR5, FHR-5complement factor H-related protein 5, FHR5MGC133240, FLJ10549
Gene Symbols CFHR5
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Complement Factor H-related 5/CFHR5 Glu19-Glu569 Accession # Q9BXR6
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81494
Test Specificity Detects human Complement Factor H-related 5/CFHR5 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.