Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Contactin-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB9041SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB9041SP 25 μg
MAB9041100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB9041SP Supplier R&D Systems Supplier No. MAB9041SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Contactin-1 Monoclonal specifically detects Contactin-1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Contactin-1
Applications Western Blot
Classification Monoclonal
Clone 2332B
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q12860
Gene Alias CNTN1, contactin 1, contactin-1, Contactin1, F11, F3, F3cam, Glycoprotein gp135, GP135, Neural cell surface protein F3
Host Species Rabbit
Immunogen Human embryonic kidney cell, HEK293-derived human Contactin-1, Met1-Ser993, Accession No. Q12860
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 1272.0
Test Specificity Detects human Contactin-1 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.