Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Contactin-2/TAG1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB17141
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB17141 100 μg
MAB17141SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB17141 Supplier R&D Systems Supplier No. MAB17141
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Contactin-2/TAG1 Monoclonal specifically detects Contactin-2/TAG1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Contactin-2/TAG1
Applications Western Blot
Classification Monoclonal
Clone 372913
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q02246
Gene Alias Axonal glycoprotein TAG-1, Axonin-1, axonin-1 cell adhesion molecule, AXT, CNTN2, contactin 2 (axonal), contactin 2 (transiently expressed), contactin-2, Contactin2, DKFZp781D102, FLJ37193, MGC157722, TAG1, TAG-1, TAX, TAX1, TAX-1, TAX1FLJ42746, Transient axonal glycoprotein 1, transiently-expressed axonal glycoprotein
Gene Symbols CNTN2
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Contactin-2/TAG1 Leu29-Asn1012 Accession # Q02246
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6900
Test Specificity Detects human Contactin-2/TAG1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) Contactin-4, rhContactin-5, recombinant mouse (rm) Contactin-2, and rmContactin-3 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.