Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Contactin-4 Isoform a Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2205
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2205 100 μg
AF2205SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2205 Supplier R&D Systems Supplier No. AF2205
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Contactin-4 Polyclonal specifically detects Contactin-4 in Human samples. It is validated for Western Blot, Neutralization.

Specifications

Antigen Contactin-4
Applications Western Blot, Neutralization
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Neutralization 5 ug/mL
Gene Accession No. Q8IWV2
Gene Alias AXCAM, axonal-associated cell adhesion molecule, BIG-2, BIG-2Brain-derived immunoglobulin superfamily protein 2, CNTN4, contactin 4, contactin-4, Contactin4, MGC33615, neural cell adhesion protein BIG-2
Gene Symbols CNTN4
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Contactin-4 isoform a Asp19-Gly1001 Accession # Q8IWV2
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 152330
Test Specificity Detects Contactin-4 isoform a in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) CNTN2, recombinant mouse CNTN3, and rhCNTN5 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.