Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Coronavirus Spike Protein Antibody (028), Alexa Fluor™ 647 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290334AF647
Description
Human Coronavirus Spike Protein Monoclonal antibody specifically detects Human Coronavirus Spike Protein in Virus samples. It is validated for ELISASpecifications
Human Coronavirus Spike Protein | |
Monoclonal | |
Alexa Fluor 647 | |
HKU1, Spike glycoprotein | |
This antibody was obtained from a rabbit immunized with purified, recombinant HCoV-HKU1 (isolate N1) Spike/S1 (YP_173238.1; Met1-Arg760) | |
0.1 mL | |
Primary | |
Virus | |
Purified |
ELISA | |
028 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
3200426 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction