Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Coronavirus Spike Protein Antibody (11), Alexa Fluor™ 750, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314653AF750
Description
Human Coronavirus Spike Protein Monoclonal antibody specifically detects Human Coronavirus Spike Protein in HCoV-229E samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
Human Coronavirus Spike Protein | |
Monoclonal | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
3200426 | |
Store at 4°C in the dark. | |
Purified |
Western Blot, ELISA, Immunofluorescence | |
11 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
HKU1, Spike glycoprotein | |
This product is a recombinant monoclonal antibody expressed from HEK293 cells. | |
0.1 mL | |
Primary | |
HCoV-229E | |
Antibody | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction