Learn More
Human COUP-TF I/NR2F1 Antibody (Clone H8132), R&D Systems™

Description
Specifications
Specifications
| Antigen | COUP-TF I/NR2F1 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Clone | H8132 |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation |
| Formulation | A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide |
| Gene Alias | COUP transcription factor 1, COUPTF I, COUP-TF1, COUP-TFI, EAR-3COUP transcription factor I, EAR3COUP-TF I, ERBAL3, ERBAL3V-erbA-related protein 3, NR2F1, Nuclear receptor subfamily 2 group F member 1, nuclear receptor subfamily 2, group F, member 1, SVP44, TCFCOUP1, TFCOUP1NR2F2, transcription factor COUP 1 (chicken ovalbumin upstream promoter 1, v-erb-ahomolog-like 3) |
| Gene Symbols | NR2F1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.