Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human COUP-TF II/NR2F2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. PPH714700
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PPH714700 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PPH714700 Supplier R&D Systems Supplier No. PPH714700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

COUP-TF II/NR2F2 Monoclonal specifically detects COUP-TF II/NR2F2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen COUP-TF II/NR2F2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone H7147
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation
Formulation A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide
Gene Alias Apolipoprotein A-I regulatory protein 1, ARP-1, ARP1ADP-ribosylation factor related protein 1, chicken ovalbumin upstream promoter transcription factor 2, COUP transcription factor 2, COUP transcription factor II, COUPTF II, COUP-TF II, COUP-TF2, COUPTFB, COUPTFII, COUP-TFII, MGC117452, NR2F2, Nuclear receptor subfamily 2 group F member 2, nuclear receptor subfamily 2, group F, member 2, SVP40, TFCOUP2apolipoprotein AI regulatory protein 1
Gene Symbols NR2F2
Host Species Mouse
Immunogen Recombinant human COUP-TF II/NR2F2 Amino Acids 43-64
Purification Method Protein A or G purified from ascites
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7026
Test Specificity This antibody specifically recognizes human COUP-TF II and cross-reacts with rat COUP-TF II. This antibody does not cross-react with human COUP-TF I or EAR2. Not yet tested in other species.
Target Species Human
Content And Storage This antibody is stable for greater than six months when held at -20degrees C in a manual defrost freezer or at -70degrees C. Upon thawing, the antibody can be stored at 2degrees - 8degrees C for at least 1 month without detectable loss of activity. Avoid repeated freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.