Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human COX-2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB4198
This item is not returnable.
View return policy
Description
COX-2 Monoclonal specifically detects COX-2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
COX-2 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunocytochemistry 8-25 ug/mL | |
P35354 | |
PTGS2 | |
E. coli-derived recombinant human COX-2 Ala18-Ser112 and Gln386-Leu604 Accession # P35354 | |
100 μg | |
Primary | |
Detects human COX-2 in Western blots. | |
Human | |
Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry | |
495222 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
COX2, COX-2, COX2cyclooxygenase 2b, cyclooxygenase-2, EC 1.14.99, EC 1.14.99.1, GRIPGHS, hCox-2, PGG/HS, PGH synthase 2, PGHS-2, PHS II, PHS-2, PHS-II, prostaglandin G/H synthase 2, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 2, Prostaglandin-endoperoxide synthase 2, prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase), PTGS2 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
5743 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction