Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CRABP2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5740
This item is not returnable.
View return policy
Description
CRABP2 Polyclonal specifically detects CRABP2 in Human samples. It is validated for Western Blot.Specifications
CRABP2 | |
Polyclonal | |
Unconjugated | |
P29373 | |
CRABP2 | |
E. coli-derived recombinant human CRABP2 Met1-Glu138 Accession # P29373 | |
100 μg | |
Primary | |
Detects endogenous human CRABP2 in Western blots. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.5 ug/mL | |
cellular retinoic acid binding protein 2, Cellular retinoic acid-binding protein II, CRABP-II, CRABP-IIcellular retinoic acid-binding protein 2, RBP6 | |
Goat | |
Affinity Purified | |
RUO | |
1382 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction