Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CRACC/SLAMF7 Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $526.50
Specifications
Antigen | CRACC/SLAMF7 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
CRACC/SLAMF7 Polyclonal specifically detects CRACC/SLAMF7 in Human samples. It is validated for Western Blot.Specifications
CRACC/SLAMF7 | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7, SLAMF7 | |
SLAMF7 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q9NQ25 | |
57823 | |
Mouse myeloma cell line NS0-derived recombinant human CRACC/SLAMF7 Lys27-Ser225 Accession # Q9NQ25 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CRACC/SLAMF7 in direct ELISAs and Western blots |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title