Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CRACC/SLAMF7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $2070.00
Specifications
| Antigen | CRACC/SLAMF7 |
|---|---|
| Clone | 235614 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB1906SP
![]() |
R&D Systems
MAB1906SP |
25 μg |
Each for $200.50
|
|
|||||
MAB1906100
![]() |
R&D Systems
MAB1906100 |
100 μg |
Each for $573.00
|
|
|||||
MAB1906500
![]() |
R&D Systems
MAB1906500 |
500 μg |
Each for $2,070.00
|
|
|||||
Description
CRACC/SLAMF7 Monoclonal specifically detects CRACC/SLAMF7 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| CRACC/SLAMF7 | |
| LYOPH | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 19A, CD2 subset 1, CD2-like receptor activating cytotoxic cells, CD319, CD319 antigen, CRACCCD2-like receptor-activating cytotoxic cells, CS119A24 protein, Membrane protein FOAP-12, Novel Ly9, novel LY9 (lymphocyte antigen 9) like protein, Protein 19A, SLAM family member 7, SLAMF7 | |
| SLAMF7 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 235614 | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Mouse | |
| Human | |
| Q9NQ25 | |
| 57823 | |
| Mouse myeloma cell line NS0-derived recombinant human CRACC/SLAMF7 Lys27-Ser225 Accession # Q9NQ25 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CRACC/SLAMF7 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human NTB-AR is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title