Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CRBN Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

$188.28 - $520.64

Specifications

Antigen CRBN
Clone 978015
Dilution Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 5-25 ug/mL
Applications Immunohistochemistry, Immunocytochemistry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB9574SP
GREENER_CHOICE
View Documents
R&D Systems
MAB9574SP
25 μg
Each for $188.28
Only null left
Add to Cart
 
MAB9574100
GREENER_CHOICE
View Documents
R&D Systems
MAB9574100
100 μg
Each for $520.64
Only null left
Add to Cart
 
Description

Description

CRBN Monoclonal specifically detects CRBN in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications

Specifications

CRBN
Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 5-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
cereblon, DKFZp781K0715, MGC27358, MRT2, MRT2A, non-syndromic, autosomal recessive, 2A, protein cereblon, protein x 0001
Synthetic peptide Human CRBN Accession # Q96SW2
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human CRBN in immunocytochemistry and immunohistochemistry.
978015
Immunohistochemistry, Immunocytochemistry
Unconjugated
Mouse
Human
Q96SW2
51185
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only (RUO)

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.