Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Creatine Kinase MM/CKMM Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems G113C
This item is not returnable.
View return policy
Description
Creatine Kinase, Muscle/CKMM Polyclonal specifically detects Creatine Kinase, Muscle/CKMM in Human samples. It is validated for .Specifications
Creatine Kinase, Muscle/CKMM | |
LYOPH | |
Hidden | |
CKM | |
Affinity Purified | |
RUO | |
1158 | |
Human | |
IgG |
Polyclonal | |
Unconjugated | |
CKM, CKMM, CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK | |
Goat | |
0.1 mg | |
Primary | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Store the unopened product at 2 - 8degrees C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction