Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human Creatine Kinase MM/CKMM Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB5564
This item is not returnable.
View return policy
Description
Creatine Kinase, Muscle/CKMM Monoclonal specifically detects Creatine Kinase, Muscle/CKMM in Human samples. It is validated for Western Blot, Simple Western.Specifications
Creatine Kinase, Muscle/CKMM | |
Monoclonal | |
LYOPH | |
Western Blot 0.1 ug/mL, Simple Western 1 ug/mL | |
CKM, CKMM, CKMMcreatine kinase M-type, Creatine Kinase Isoenzyme, Creatine kinase M chain, creatine kinase, muscle, creatine kinase-M, EC 2.7.3, EC 2.7.3.2, M-CK | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1158 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Western Blot | |
492731 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CKM | |
Human cardiac muscle-derived Creatine Kinase, Muscle/CKMM | |
100 μg | |
Primary | |
Detects human Creatine Kinase, Muscle/CKMM in direct ELISAs and Western blots. | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction