Learn More
Human CREB Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody
Supplier:  R&D Systems AF2989SP
Catalog No. AF2989SP
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
CREB Polyclonal specifically detects CREB in Human samples. It is validated for Western Blot, Immunohistochemistry, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.
Specifications
Specifications
CREB
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
active transcription factor CREB, cAMP responsive element binding protein 1, cAMP-response element-binding protein-1, cAMP-responsive element-binding protein 1, CREB, CREB1, CREB-1, cyclic AMP-responsive element-binding protein 1, MGC9284, transactivator protein
Goat
Affinity Purified
RUO
1385
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
ChIP Assay, Immunocytochemistry, Immunohistochemistry
LYOPH
Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/5 x 10^6 cells, Immunocytochemistry 5-15 ug/mL
P16220
CREB1
E. coli-derived recombinant human CREB Lys136-Asp341 Accession # P16220
25 μg
Primary
Detects human CREB in Western blots.
Human
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only