Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CREG Antibody, R&D Systems™

Goat Polyclonal Antibody
$200.50 - $549.50
Specifications
Antigen | CREG |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
CREG Polyclonal specifically detects CREG in Human samples. It is validated for Western Blot.Specifications
CREG | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
cellular repressor of E1A-stimulated genes, cellular repressor of E1A-stimulated genes 1CREG, CREG1, protein CREG1 | |
CREG1 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
O75629 | |
8804 | |
Mouse myeloma cell line NS0-derived recombinant human CREG Arg32-Gln220 Accession # O75629 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human CREG in direct ELISAs and Western blots. In Western blots, approximately 50% cross-reactivity with recombinant mouse CREG is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title