Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CRELD1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3128
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3128 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB3128 Supplier R&D Systems Supplier No. MAB3128
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CRELD1 Monoclonal specifically detects CRELD1 in Human samples. It is validated for Neutralization.

Specifications

Antigen CRELD1
Applications Neutralization
Classification Monoclonal
Clone 842314
Concentration LYOPH
Conjugate Unconjugated
Dilution Neutralization 0.15-0.75 ug/mL
Gene Accession No. Q96HD1
Gene Alias atrioventricular septal defect 2, AVSD2, CIRRIN, cysteine-rich with EGF-like domain protein 1, cysteine-rich with EGF-like domains 1, DKFZp566D213
Gene Symbols CRELD1
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CRELD-1 Gln30-Glu362 Accession # Q96HD1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 78987
Test Specificity Detects human CRELD-1 in ELISA.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.