Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CRELD2 alpha Isoform Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF3555
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF3555 100 μg
AF3555SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF3555 Supplier R&D Systems Supplier No. AF3555
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CRELD2 Polyclonal specifically detects CRELD2 in Human samples. It is validated for Western Blot.

Specifications

Antigen CRELD2
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. AAH50675
Gene Alias cysteine-rich with EGF-like domain protein 2, cysteine-rich with EGF-like domains 2, DKFZp667O055, MGC11256
Gene Symbols CRELD2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CRELD2 alpha Isoform Ala21-Leu353 Accession # AAH50675
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79174
Test Specificity Detects human CRELD2 alpha isoform in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 30% cross-reactivity with recombinant mouse (rm) CRELD2 is observed and less than 1% cross-reactivity with recombinant human CRELD1 and rmCRELD1 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.