Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human CRELD2 ELISA Kit

Catalog No. p-7219305
Encompass_Preferred
Click to view available options
Quantity:
5 x 96 Tests
96 Tests

ELISA

Human CRELD2 quantitates human CRELD2 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CRELD2.

CRELD2 may regulate transport of alpha4-beta2 neuronal acetylcholine receptor.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 5730592L21Rik; C85758; CREL2; Creld2; cysteine rich with EGF like domains 2; cysteine rich with EGF-like domains 2; cysteine-rich with EGF-like domain protein 2; cysteine-rich with EGF-like domains 2; DKFZp667O055; MGC11256; Protein disulfide isomerase CRELD2; RGD1310614; UNQ185/PRO211
Assay Range 0.137 to 100 ng/mL
Assay Sensitivity 0.13 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 79174
Gene Symbol CRELD2
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target CRELD2
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.