Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CRIM1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF1917SP
This item is not returnable.
View return policy
Description
CRIM1 Polyclonal specifically detects CRIM1 in Human samples. It is validated for Western Blot.Specifications
CRIM1 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CRIM-1, cysteine rich transmembrane BMP regulator 1 (chordin-like), cysteine-rich motor neuron 1, cysteine-rich motor neuron 1 protein, Cysteine-rich repeat-containing protein S52, MGC138194, S52 | |
Goat | |
Affinity Purified | |
RUO | |
51232 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Q9NZV1 | |
CRIM1 | |
Mouse myeloma cell line NS0-derived recombinant human CRIM1 Leu35-Ser939 Accession # Q9NZV1 | |
25 μg | |
Primary | |
Detects human CRIM1 in direct ELISAs and Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction