Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Human CRIM1 ELISA Kit
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
Description
Human CRIM1 quantitates human CRIM1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human CRIM1.
CRIM1 (cysteine-rich motor neuron 1), a glycosylated type I transmembrane protein, plays a role in tissue development i.e. capillary formation and maintenance during angiogenesis. It contains an N-terminal IGF-binding protein-like motif and six von Willebrand-like cysteine-rich repeats (CRRs) in its extracellular domain. CRIM1 interacts with BMP4 and BMP7 via the CRRs and functions as an antagonist. CRIM1 is developmentally expressed in a number of tissues including the pancreas, kidney, placenta, brain and blood vessels. CRIM1 may participate in CNS and placental development by interacting with growth factors involved in motor neuron differentiation and survival.
Specifications
Specifications
| Accession Number | AU015004; Crim1; CRIM-1; cysteine rich transmembrane BMP regulator 1; cysteine rich transmembrane BMP regulator 1 (chordin like); cysteine rich transmembrane BMP regulator 1 (chordin-like); cysteine-rich motor neuron 1; cysteine-rich motor neuron 1 protein; Cysteine-rich repeat-containing protein S52; Processed cysteine-rich motor neuron 1 protein; S52; UNQ1886/PRO4330 |
| Assay Range | 0.041 to 10 ng/mL |
| Assay Sensitivity | 0.041 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 51232 |
| Gene Symbol | CRIM1 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction