Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Cripto-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF145
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF145 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF145 Supplier R&D Systems Supplier No. AF145
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CRIPTO Polyclonal specifically detects CRIPTO in Human samples. It is validated for Western Blot.

Specifications

Antigen CRIPTO
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL
Gene Accession No. P13385
Gene Alias CFC-2, CR, Cripto-1, Cripto-1 growth factor, CRIPTOCRGF, Epidermal growth factor-like cripto protein CR1, TDGF1, TDGF-1, teratocarcinoma-derived growth factor 1
Gene Symbols TDGF1
Host Species Goat
Immunogen E. coli-derived recombinant human Cripto-1 Arg38-Tyr188 Accession # P13385
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6997
Test Specificity Detects human Cripto-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse Cripto is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.