Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CRISP-3 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems IC23972V100UG
This item is not returnable.
View return policy
Description
CRISP-3 Monoclonal specifically detects CRISP-3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
CRISP-3 | |
Monoclonal | |
0.2 mg/mL | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
P54108.1 | |
CRISP3 | |
Mouse myeloma cell line NS0-derived recombinant human CRISP-3 Asn21-Tyr245 (Ser106Pro & Ala134Ser) Accession # P54108.1 | |
RUO | |
10321 | |
Human | |
IgG2b |
Flow Cytometry | |
295208 | |
Alexa Fluor 405 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
Aeg2, CRISP3, CRISP-3MGC126588, CRS3, cysteine-rich secretory protein 3, cysteine-rich secretory protein-3, dJ442L6.3, SGP28, SGP28Aeg2, specific granule protein (28 kDa), Specific granule protein of 28 kDa | |
Mouse | |
100 μg | |
Primary | |
Detects human CRISP-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CRISP-2 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction