Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CRISP-3 Alexa Fluor 594-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC23972T100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC23972T100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC23972T100 Supplier R&D Systems Supplier No. IC23972T100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CRISP-3 Monoclonal specifically detects CRISP-3 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen CRISP-3
Applications Flow Cytometry
Classification Monoclonal
Clone 295208
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 594
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. P54108.1
Gene Alias Aeg2, CRISP3, CRISP-3MGC126588, CRS3, cysteine-rich secretory protein 3, cysteine-rich secretory protein-3, dJ442L6.3, SGP28, SGP28Aeg2, specific granule protein (28 kDa), Specific granule protein of 28 kDa
Gene Symbols CRISP3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CRISP-3 Asn21-Tyr245 (Ser106Pro & Ala134Ser) Accession # P54108.1
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10321
Test Specificity Detects human CRISP-3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CRISP-2 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2b
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.