Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CRISP-3 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems BAF2397
This item is not returnable.
View return policy
Description
CRISP-3 Polyclonal specifically detects CRISP-3 in Human samples. It is validated for Western Blot.Specifications
CRISP-3 | |
Polyclonal | |
Biotin | |
CAA63984 | |
CRISP3 | |
Mouse myeloma cell line NS0-derived recombinant human CRISP-3, Asn21-Tyr245 (Ser134Ala), Accession # CAA63984 | |
50 μg | |
Primary | |
Detects human CRISP-3 in Western blots. In Western blots, approximately 15% cross-reactivity with recombinant human CRISP-2 is observed. | |
Human | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 0.1 ug/mL | |
Aeg2, CRISP3, CRISP-3MGC126588, CRS3, cysteine-rich secretory protein 3, cysteine-rich secretory protein-3, dJ442L6.3, SGP28, SGP28Aeg2, specific granule protein (28 kDa), Specific granule protein of 28 kDa | |
Goat | |
Affinity Purified | |
RUO | |
10321 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction