Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CRTAC1 Isoform 1 Biotinylated Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems BAF5234
This item is not returnable.
View return policy
Description
CRTAC1 Polyclonal specifically detects CRTAC1 in Human samples. It is validated for Western Blot.Specifications
| CRTAC1 | |
| Polyclonal | |
| Biotin | |
| Q9NQ79 | |
| CRTAC1 | |
| Mouse myeloma cell line NS0-derived recombinant human CRTAC1 isoform 1 Ser28-Cys661 Accession # Q9NQ79 | |
| 50 μg | |
| Primary | |
| Detects human CRTAC1 Isoform 1 in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human CRTAC1 Isoform 2 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| ASPIC, ASPIC1, ASPIC1CEP-68acidic secreted protein in cartilage, cartilage acidic protein 1, CEP68, chondrocyte expressed protein 68 kDa CEP-68,68 kDa chondrocyte-expressed protein, FLJ10320 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 55118 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction