Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CRTAM Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF1695
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF1695 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF1695 Supplier R&D Systems Supplier No. AF1695
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

CRTAM Polyclonal specifically detects CRTAM in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen CRTAM
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 0.2-0.8 ug/mL
Gene Accession No. O95727
Gene Alias CD355, CD355 antigen, class I MHC restricted T cell associated molecule, class-I MHC-restricted T cell associated molecule, Class-I MHC-restricted T-cell-associated molecule, cytotoxic and regulatory T cell molecule, cytotoxic and regulatory T-cell molecule
Gene Symbols CRTAM
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human CRTAM Ser18-Ser286 Accession # O95727
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56253
Test Specificity Detects human CRTAM in ELISAs and Western blots. In sandwich immunoassays, less than 2% cross-reactivity with recombinant mouse CRTAM is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.