Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CRTH-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB3338SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3338SP 25 μg
MAB3338 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB3338SP Supplier R&D Systems Supplier No. MAB3338SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

CRTH-2/GPR44 Monoclonal specifically detects CRTH-2/GPR44 in Human samples. It is validated for Flow Cytometry, CyTOF-ready.

Specifications

Antigen CRTH-2/GPR44
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 301108
Concentration LYOPH
Conjugate Unconjugated
Dilution Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q9Y5Y4
Gene Alias CD294, CD294 antigen, Chemoattractant receptor-homologous molecule expressed on Th2 cells, CRTH2, CRTH2chemoattractant receptor homologous molecule expressed on T helper type 2 cells, DL1R, DP2, G protein-coupled receptor 44, GPR44, PGD2R, PTGDR2, putative G-protein coupled receptor 44
Gene Symbols PTGDR2
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human CRTH-2 Met1-Ser395 Accession # Q9Y5Y4
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11251
Test Specificity Detects human CRTH-2. Stains human CRTH-2-transfected cells but not parental cells.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.