Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CSRP2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF6196SP
This item is not returnable.
View return policy
Description
CSRP2 Polyclonal specifically detects CSRP2 in Human samples. It is validated for Western Blot.Specifications
CSRP2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CRP2, CRP2LIM domain only protein 5, cysteine and glycine-rich protein 2, Cysteine-rich protein 2, LMO5LMO-5, SMLIM, SmLIMLIM domain only 5, smooth muscle, Smooth muscle cell LIM protein | |
Goat | |
Affinity Purified | |
RUO | |
1466 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 2 ug/mL | |
Q16527 | |
CSRP2 | |
E. coli-derived recombinant human CSRP2 Met1-Gln193 Accession # Q16527 | |
25 μg | |
Primary | |
Detects human CSRP2 in Western blots. | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction