Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human CTGF/CCN2 C-Terminus Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF660
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF660 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF660 Supplier R&D Systems Supplier No. AF660
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 3 publications

CTGF/CCN2 Polyclonal specifically detects CTGF/CCN2 in Human samples. It is validated for Immunohistochemistry.

Specifications

Antigen CTGF/CCN2
Applications Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Immunohistochemistry 5-15 ug/mL
Gene Accession No. P29279
Gene Alias CCN2, CCN2IGFBP-8, connective tissue growth factor, CTGRP, Fisp12, HCS24, Hypertrophic chondrocyte-specific protein 24, IBP-8, IGF-binding protein 8, IGFBP-8, IGFBP8CCN family member 2, Insulin-like growth factor-binding protein 8, MGC102839, NOV2
Gene Symbols CTGF
Host Species Goat
Immunogen E. coli-derived recombinant human CTGF/CCN2 C-Terminus Glu247-Ala349 Accession # P29279
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1490
Test Specificity Detects human CTGF/CCN2 C-Terminus in direct ELISAs. In direct ELISAs, approximately 15% cross-reactivity with recombinant rat CTGF is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.