Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CTLA-4 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
$227.50 - $502.50
Specifications
Antigen | CTLA-4 |
---|---|
Clone | 2188B |
Dilution | Flow Cytometry 0.25 μg/106 cells, Immunocytochemistry 8-25 μg;/mL, CyTOF-ready |
Applications | Flow Cytometry, Immunocytochemistry, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB3253SP
![]() |
R&D Systems
MAB3253SP |
25 μg |
Each for $227.50
|
|
|||||
MAB3253100
![]() |
R&D Systems
MAB3253100 |
100 μg |
Each for $502.50
|
|
|||||
Description
CTLA-4 Monoclonal specifically detects CTLA-4 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
CTLA-4 | |
Flow Cytometry 0.25 μg/106 cells, Immunocytochemistry 8-25 μg;/mL, CyTOF-ready | |
Monoclonal | |
Rabbit | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4 | |
Chinese hamster ovary cell line CHO-derived recombinant human CTLA-4, Ala37-Phe162, Accession No. P16410 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CTLA-4 in direct ELISAs. |
2188B | |
Flow Cytometry, Immunocytochemistry, CyTOF | |
Unconjugated | |
Human | |
P16410 | |
1493.0 | |
IgG | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title