Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human CTRP1/C1qTNF1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $468.00
Specifications
Antigen | C1qTNF1/CTRP1 |
---|---|
Clone | 395526 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
C1qTNF1/CTRP1 Monoclonal specifically detects C1qTNF1/CTRP1 in Human samples. It is validated for Western Blot.Specifications
C1qTNF1/CTRP1 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9BXJ1 | |
114897 | |
Mouse myeloma cell line NS0-derived recombinant human CTRP1/C1qTNF1 Arg26-Pro281 Accession # Q9BXJ1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects human CTRP1/C1qTNF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CTRP4, rhCTRP5, or rhC1qL2 is observed. |
395526 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C1q and tumor necrosis factor related protein 1, C1qTNF1, CTRP1G protein-coupled receptor-interacting protein, FLJ90694, G protein coupled receptor interacting protein, GIPcomplement C1q tumor necrosis factor-related protein 1, ZSIG37 | |
C1QTNF1 | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title